5-Chloro-2-methoxyisonicotinic acid ≥95%

Supplier: Matrix Scientific

073843-250MG 073843-1G 073843-5G
102533-286EA 684.62 USD
102533-286 102533-288 102533-290
5-Chloro-2-methoxyisonicotinic acid ≥95%
5-Chloro-2-methoxyisonicotinic acid

Hazard: Irritant

Formula: C7H6ClNO3
CAS Number: 88912-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR