2-Acetamido-3-bromo-5-fluoropyridine ≥95%

Supplier: Matrix Scientific

073842-1G 073842-5G
102533-282EA 569.23 USD
102533-282 102533-284
2-Acetamido-3-bromo-5-fluoropyridine ≥95%
2-Acetamido-3-bromo-5-fluoropyridine

Hazard: Irritant

Formula: C₇H₆BrFN₂O
MDL Number: MFCD11053869
CAS Number: 1065074-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR