6-Chlorobenzo[d]isoxazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

073839-250MG 073839-1G
102533-266EA 275.38 USD
102533-266 102533-268
6-Chlorobenzo[d]isoxazole-3-carboxylic acid ≥95%
6-Chlorobenzo[d]isoxazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₄ClNO₃
MDL Number: MFCD09027540
CAS Number: 28691-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR