tert-Butyl-4-(bromomethyl)benzylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(bromomethyl)benzylcarbamate, N-BOC-4-(Bromomethyl)benzylamine

073837-1G 073837-5G
102533-258EA 341.54 USD
102533-258 102533-260
tert-Butyl-4-(bromomethyl)benzylcarbamate ≥95%
tert-Butyl-4-(bromomethyl)benzylcarbamate

Hazard: Irritant

Formula: C₁₃H₁₈BrNO₂
MW: 300.1900 g/mol
MDL Number: MFCD11974929
CAS Number: 187283-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR