3-Oxocyclopentanecarboxylic acid ≥95%

Supplier: Matrix Scientific

073835-1G 073835-5G
102533-250EA 184.62 USD
102533-250 102533-252
3-Oxocyclopentanecarboxylic acid ≥95%
3-Oxocyclopentanecarboxylic acid

Hazard: Irritant

Formula: C6H8O3
MDL Number: MFCD01320173
CAS Number: 98-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR