tert-Butyl-4-(2-oxo-2,4-dihydro-1H-benzo[d][1,3]oxazin-1-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

073834-250MG 073834-1G
102533-246EA 924.62 USD
102533-246 102533-248
tert-Butyl-4-(2-oxo-2,4-dihydro-1H-benzo[d][1,3]oxazin-1-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(2-oxo-2,4-dihydro-1H-benzo[d][1,3]oxazin-1-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₄
CAS Number: 162045-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR