3,6-Dibromopyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3,6-Dibromo-2-pyrazinecarboxylic Acid

073833-250MG 073833-1G 073833-5G
102533-240EA 640 USD
102533-240 102533-242 102533-244
3,6-Dibromopyrazine-2-carboxylic acid ≥95%
3,6-Dibromopyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C5H2Br2N2O2
MW: 281.89 g/mol
MDL Number: MFCD13193674
CAS Number: 957230-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR