5-Bromo-N-methylpyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

073832-250MG 073832-1G 073832-5G
102533-234EA 684.62 USD
102533-234 102533-236 102533-238
5-Bromo-N-methylpyrimidin-4-amine ≥95%
5-Bromo-N-methylpyrimidin-4-amine

Hazard: Irritant

Formula: C₅H₆BrN₃
MW: 188.03 g/mol
MDL Number: MFCD03861147
CAS Number: 56181-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR