4-Chloro-N-methylpyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

073830-1G 073830-5G
102533-224EA 553.85 USD
102533-224 102533-226
4-Chloro-N-methylpyrimidin-2-amine ≥95%
4-Chloro-N-methylpyrimidin-2-amine

Hazard: Irritant

Formula: C₅H₆ClN₃
MW: 143.5700 g/mol
MDL Number: MFCD09834973
CAS Number: 22404-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR