5-Bromo-4-chloropyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

073826-1G 073826-5G
102533-202EA 250.77 USD
102533-202 102533-204
5-Bromo-4-chloropyrimidin-2-amine ≥95%
5-Bromo-4-chloropyrimidin-2-amine

Hazard: Irritant

Formula: C₄H₃BrClN₃
MDL Number: MFCD11046979
CAS Number: 1044767-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR