Methyl-2-cyanoisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-cyanoisonicotinate

073823-1G 073823-5G
102533-184EA 186.15 USD
102533-184 102533-186
Methyl-2-cyanoisonicotinate ≥95%
Methyl-2-cyanoisonicotinate

Hazard: Toxic

Formula: C₈H₆N₂O₂
MW: 162.148 g/mol
MDL Number: MFCD07367894
CAS Number: 94413-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR