5-Bromo-2-chloro-4-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073821-250MG 073821-1G
102533-176EA 213.85 USD
102533-176 102533-178
5-Bromo-2-chloro-4-methylpyrimidine ≥95%
5-Bromo-2-chloro-4-methylpyrimidine

Hazard: Irritant

Formula: C₅H₄BrClN₂
MW: 207.4557 g/mol
MDL Number: MFCD13193654
CAS Number: 633328-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR