tert-Butyl (2-bromothiazol-5-yl)carbamate ≥95%

Supplier: Matrix Scientific

073818-250MG 073818-1G 073818-5G
102533-158EA 867.69 USD
102533-158 102533-160 102533-162
tert-Butyl (2-bromothiazol-5-yl)carbamate ≥95%
tert-Butyl (2-bromothiazol-5-yl)carbamate

Hazard: Irritant

Formula: C₈H₁₁BrN₂O₂S
MW: 279.1541 g/mol
MDL Number: MFCD13195442
CAS Number: 1094070-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR