3-Bromo-1H-pyrazolo[3,4-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073815-250MG 073815-1G
102533-142EA 967.69 USD
102533-142 102533-144
3-Bromo-1H-pyrazolo[3,4-d]pyrimidine ≥95%
3-Bromo-1H-pyrazolo[3,4-d]pyrimidine

Hazard: Irritant

Formula: C5H3BrN4
MW: 199.0100 g/mol
MDL Number: MFCD13193631
CAS Number: 1251033-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR