Ethyl-3-(pyridin-2-ylamino)propanoate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2-Pyridylamino)propionic acid ethyl ester, Ethyl 3-(2-Pyridylamino)propionate, Ethyl 3-(pyridin-2-ylamino)propanoate

073814-1G 073814-5G
102533-138EA 38.46 USD
102533-138 102533-140
Ethyl-3-(pyridin-2-ylamino)propanoate ≥95%
Ethyl-3-(pyridin-2-ylamino)propanoate

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MW: 194.2300 g/mol
MDL Number: MFCD09833621
CAS Number: 103041-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR