Methyl-2-bromoimidazo[1,2-a]pyridine-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-bromoimidazo[1,2-a]pyridine-6-carboxylate

073812-250MG 073812-1G 073812-5G
102533-126EA 640 USD
102533-126 102533-128 102533-130
Methyl-2-bromoimidazo[1,2-a]pyridine-6-carboxylate ≥95%
Methyl 2-bromoimidazo[1,2-a]pyridine-6-carboxylate

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MW: 255.0700 g/mol
MDL Number: MFCD13195435
CAS Number: 1042141-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR