2-Methoxypyrimidine-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

073811-250MG 073811-1G 073811-5G
102533-120EA 640 USD
102533-120 102533-122 102533-124
2-Methoxypyrimidine-4-carbaldehyde ≥95%
2-Methoxypyrimidine-4-carbaldehyde

Hazard: Irritant

Formula: C₆H₆N₂O₂
CAS Number: 164738-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR