2,4-Dichloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073810-250MG 073810-1G 073810-5G
102533-114EA 355.38 USD
102533-114 102533-116 102533-118
2,4-Dichloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%
2,4-Dichloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₁₃H₉Cl₂N₃O₂S
MW: 342.2000 g/mol
MDL Number: MFCD13193624
CAS Number: 934524-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR