Benzo[d]oxazol-2-ylmethanol ≥95%

Supplier: Matrix Scientific

073808-250MG 073808-1G 073808-5G
102533-102EA 355.38 USD
102533-102 102533-104 102533-106
Benzo[d]oxazol-2-ylmethanol ≥95%
Benzo[d]oxazol-2-ylmethanol

Hazard: Irritant

Formula: C₈H₇NO₂
MW: 149.1467 g/mol
MDL Number: MFCD07368333
CAS Number: 77186-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR