Benzo[d]thiazole-7-carboxylic acid ≥95%
Supplier: Matrix Scientific
073806-250MG
073806-1G
102533-094EA
378.96
USD
102533-094
102533-096
Benzo[d]thiazole-7-carboxylic acid ≥95%
Benzo[d]thiazole-7-carboxylic acid
Hazard: Irritant
Formula:
C₈H₅NO₂S MW: 179.1958 g/mol |
MDL Number:
MFCD13195419 CAS Number: 677304-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...