2-Aminobenzo[d]thiazole-7-carboxylic acid ≥95%

Supplier: Matrix Scientific

073805-250MG 073805-1G
102533-090EA 344.62 USD
102533-090 102533-092
2-Aminobenzo[d]thiazole-7-carboxylic acid ≥95%
2-Aminobenzo[d]thiazole-7-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂S
MW: 194.2104 g/mol
MDL Number: MFCD13195418
CAS Number: 71224-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR