Methyl-2-aminobenzo[d]thiazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-aminobenzo[d]thiazole-7-carboxylate

073804-250MG 073804-1G 073804-5G
102533-084EA 461.54 USD
102533-084 102533-086 102533-088
Methyl-2-aminobenzo[d]thiazole-7-carboxylate ≥95%
Methyl-2-aminobenzo[d]thiazole-7-carboxylate

Hazard: Irritant/Store in Freezer

Formula: C₉H₈N₂O₂S
MDL Number: MFCD09263762
CAS Number: 209459-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR