Spiro[indoline-3,4'-piperidin]-2-one ≥95%

Supplier: Matrix Scientific

073803-250MG 073803-1G
102533-080EA 826.15 USD
102533-080 102533-082
Spiro[indoline-3,4'-piperidin]-2-one ≥95%
Spiro[indoline-3,4'-piperidin]-2-one

Hazard: Irritant

Formula: C₁₂H₁₄N₂O
MW: 202.2500 g/mol
MDL Number: MFCD12198581
CAS Number: 252882-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR