3-Bromo-1H-pyrazolo[3,4-b]pyridin-5-amine ≥95%
Supplier: Matrix Scientific
073802-250MG
073802-1G
073802-5G
102533-074EA
498.46
USD
102533-074
102533-076
102533-078
3-Bromo-1H-pyrazolo[3,4-b]pyridin-5-amine ≥95%
3-Bromo-1H-pyrazolo[3,4-b]pyridin-5-amine
Hazard: Irritant
Formula:
C6H5BrN4 MW: 213.0400 g/mol |
MDL Number:
MFCD13193613 CAS Number: 1186608-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...