1-Methyl-2-(methylthio)-4,5-dihydro-1H-imidazole hydroiodide ≥95%

Supplier: Matrix Scientific

073798-250MG 073798-1G 073798-5G
102533-052EA 640 USD
102533-052 102533-054 102533-056
1-Methyl-2-(methylthio)-4,5-dihydro-1H-imidazole hydroiodide ≥95%
1-Methyl-2-(methylthio)-4,5-dihydro-1H-imidazole hydroiodide

Hazard: Irritant

Formula: C₅H₁₁IN₂S
CAS Number: 61076-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR