3-Oxabicyclo[3.1.0]hexane-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

073797-250MG 073797-1G 073797-5G
102533-046EA 684.62 USD
102533-046 102533-048 102533-050
3-Oxabicyclo[3.1.0]hexane-6-carboxylic acid ≥95%
3-Oxabicyclo[3.1.0]hexane-6-carboxylic acid

Hazard: Irritant

Formula: C6H8O3
MW: 128.13 g/mol
MDL Number: MFCD13193581
CAS Number: 693248-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR