4-Methyltetrahydro-2H-pyran-4-ol ≥95%

Supplier: Matrix Scientific

073796-250MG 073796-1G 073796-5G
102533-040EA 355.38 USD
102533-040 102533-042 102533-044
4-Methyltetrahydro-2H-pyran-4-ol ≥95%
4-Methyltetrahydro-2H-pyran-4-ol

Hazard: Irritant

Formula: C₆H₁₂O₂
MDL Number: MFCD13193576
CAS Number: 7525-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR