6-Bromospiro[chroman-2,4'-piperidin]-4-one ≥95%
Supplier: Matrix Scientific
073794-250MG
073794-1G
102533-032EA
569.23
USD
102533-032
102533-034
6-Bromospiro[chroman-2,4'-piperidin]-4-one ≥95%
6-Bromospiro[chroman-2,4'-piperidin]-4-one
Hazard: Irritant
Formula:
C₁₃H₁₄BrNO₂ MW: 296.16 g/mol |
MDL Number:
MFCD09262414 CAS Number: 690632-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...