tert-Butyl-6-bromo-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 6-bromo-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate
073793-250MG
073793-1G
073793-5G
102533-026EA
684.62
USD
102533-026
102533-028
102533-030
tert-Butyl-6-bromo-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate ≥95%
tert-Butyl-6-bromo-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate
Hazard: Irritant
Formula:
C₁₈H₂₂BrNO₄ MW: 396.2800 g/mol |
MDL Number:
MFCD05865132 CAS Number: 690632-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...