1'H-Spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one ≥95%

Supplier: Matrix Scientific

073792-250MG 073792-1G 073792-5G
102533-020EA 640 USD
102533-020 102533-022 102533-024
1'H-Spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one ≥95%
1'H-Spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
CAS Number: 202826-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR