2-Chloro-4-fluoro-3-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

073789-250MG 073789-1G 073789-5G
102533-004EA 350.81 USD
102533-004 102533-006 102533-008
2-Chloro-4-fluoro-3-methylbenzonitrile ≥95%
2-Chloro-4-fluoro-3-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₅ClFN
MDL Number: MFCD13193544
CAS Number: 796600-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR