(R)-2,3-Diaminopropanoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-D-alanine Hydrochloride, H-D-Dap-OH.HCl, H-D-Dap-OH·HCl

073788-1G 073788-5G 073788-10G
102532-002EA 143.08 USD
102532-002 102533-000 102533-002
(R)-2,3-Diaminopropanoic acid hydrochloride ≥95%
(R)-2,3-Diaminopropanoic acid hydrochloride

Hazard: Irritant

Formula: C₃H₉ClN₂O₂
MW: 140.5688 g/mol
MDL Number: MFCD00070606
CAS Number: 6018-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR