5-Bromo-4-chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

073786-250MG 073786-1G 073786-5G
102531-990EA 783.08 USD
102531-990 102531-992 102531-994
5-Bromo-4-chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine ≥95%
5-Bromo-4-chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₃BrClIN₂
MW: 357.37 g/mol
MDL Number: MFCD13193525
CAS Number: 1092579-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR