6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-[1,2,4]triazolo[1,5-a]pyridin-2-amine

073783-250MG 073783-1G 073783-5G
102531-976EA 498.46 USD
102531-976 102531-978 102531-980
6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%
6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine

Hazard: Irritant

Formula: C₆H₅BrN₄
MW: 213.0400 g/mol
MDL Number: MFCD11656619
CAS Number: 947248-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR