3-Methylimidazo[2,1-b]thiazole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

073782-250MG 073782-1G
102531-972EA 967.69 USD
102531-972 102531-974
3-Methylimidazo[2,1-b]thiazole-2-carboxylic acid ≥95%
3-Methylimidazo[2,1-b]thiazole-2-carboxylic acid

Hazard: Irritant

Formula: C7H6N2O2S
MW: 182.2000 g/mol
MDL Number: MFCD11889936
CAS Number: 1017273-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR