1-Boc-4-(5-methyl-1,3,4-oxadiazol-2-yl)piperidine ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-4-(5-Methyl-1,3,4-oxadiazol-2-yl)piperidine
073780-250MG
073780-1G
102531-962EA
967.69
USD
102531-962
102531-964
1-Boc-4-(5-methyl-1,3,4-oxadiazol-2-yl)piperidine ≥95%
1-Boc-4-(5-methyl-1,3,4-oxadiazol-2-yl)piperidine
Hazard: Irritant
Formula:
C₁₃H₂₁N₃O₃ MW: 267.3200 g/mol |
MDL Number:
MFCD13193509 CAS Number: 280110-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...