1-Phenyl-1H-imidazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

073779-250MG 073779-1G
102531-958EA 1410.77 USD
102531-958 102531-960
1-Phenyl-1H-imidazole-4-carboxylic acid ≥95%
1-Phenyl-1H-imidazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MW: 188.180 g/mol
MDL Number: MFCD13193504
CAS Number: 18075-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR