Ethyl benzo[d]isoxazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2-Benzisoxazole-3-carboxylic acid ethyl ester, 3-Ethoxycarbonyl-1,2-benzisoxazole, Ethyl 1,2-Benzisoxazole-3-carboxylate

073777-250MG 073777-1G 073777-5G
102531-946EA 569.23 USD
102531-946 102531-948 102531-950
Ethyl benzo[d]isoxazole-3-carboxylate ≥95%
Ethyl benzo[d]isoxazole-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₃
MW: 191.1834 g/mol
MDL Number: MFCD06659787
CAS Number: 57764-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR