4-Bromobenzo[d]isoxazole ≥95%

Supplier: Matrix Scientific

073776-250MG 073776-1G 073776-5G
102531-940EA 881.54 USD
102531-940 102531-942 102531-944
4-Bromobenzo[d]isoxazole ≥95%
4-Bromobenzo[d]isoxazole

Hazard: Irritant

Formula: C₇H₄BrNO
MW: 198.0168 g/mol
MDL Number: MFCD09027766
CAS Number: 1126848-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR