1-Boc-4-(3-iodo-1H-indazol-4-yl)piperazine ≥95%

Supplier: Matrix Scientific

073775-250MG 073775-1G 073775-5G
102531-934EA 967.69 USD
102531-934 102531-936 102531-938
1-Boc-4-(3-iodo-1H-indazol-4-yl)piperazine ≥95%
1-Boc-4-(3-iodo-1H-indazol-4-yl)piperazine

Hazard: Irritant

Formula: C₁₆H₂₁IN₄O₂
CAS Number: 744219-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR