(3-(4-Methylpiperazin-1-yl)phenyl)methanol ≥95%

Supplier: Matrix Scientific

073774-250MG 073774-1G 073774-5G
102531-928EA 640 USD
102531-928 102531-930 102531-932
(3-(4-Methylpiperazin-1-yl)phenyl)methanol ≥95%
(3-(4-Methylpiperazin-1-yl)phenyl)methanol

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MW: 206.2841 g/mol
MDL Number: MFCD09757582
CAS Number: 123987-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR