Pyrido[2,3-b]pyrazin-6(5H)-one ≥95%

Supplier: Matrix Scientific

073771-250MG 073771-1G 073771-5G
102531-912EA 740 USD
102531-912 102531-914 102531-916
Pyrido[2,3-b]pyrazin-6(5H)-one ≥95%
Pyrido[2,3-b]pyrazin-6(5H)-one

Hazard: Irritant

Formula: C7H5N3O
MW: 147.1300 g/mol
MDL Number: MFCD13193474
CAS Number: 35808-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR