Imidazo[1,2-a]pyrimidin-2-ylmethanamine ≥95%

Supplier: Matrix Scientific

073769-250MG 073769-1G 073769-5G
102531-902EA 355.38 USD
102531-902 102531-904 102531-906
Imidazo[1,2-a]pyrimidin-2-ylmethanamine ≥95%
Imidazo[1,2-a]pyrimidin-2-ylmethanamine

Hazard: Irritant

Formula: C₇H₈N₄
MW: 148.1700 g/mol
MDL Number: MFCD06660691
CAS Number: 843609-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR