2,6-Dichloro-3-cyanopyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-dichloropyrazine-2-carbonitrile

073768-250MG 073768-1G 073768-5G
102531-896EA 710.77 USD
102531-896 102531-898 102531-900
2,6-Dichloro-3-cyanopyrazine ≥95%
2,6-Dichloro-3-cyanopyrazine

Hazard: Irritant/Store in Freezer

Formula: C₅HCl₂N₃
MW: 173.9875 g/mol
MDL Number: MFCD13193460
CAS Number: 313339-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR