Ethyl-2-bromo-5-oxazolecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromooxazole-5-carboxylate

073767-250MG 073767-1G
102531-892EA 248.46 USD
102531-892 102531-894
Ethyl-2-bromo-5-oxazolecarboxylate ≥95%
Ethyl 2-bromo-5-oxazolecarboxylate

Hazard: Irritant

Formula: C₆H₆BrNO₃
MW: 220.0207 g/mol
MDL Number: MFCD09743496
CAS Number: 1060816-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR