5-Bromopyrimidin-4(3H)-one sulfate(2:1) ≥95%

Supplier: Matrix Scientific

073766-1G 073766-5G 073766-10G
102531-886EA 427.69 USD
102531-886 102531-888 102531-890
5-Bromopyrimidin-4(3H)-one sulfate(2:1) ≥95%
5-Bromopyrimidin-4(3H)-one sulfate(2:1)

Hazard: Irritant

Formula: C₈H₈Br₂N₄O₆S
MW: 448.0500 g/mol
MDL Number: MFCD13195360
CAS Number: 97234-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR