7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

073763-1G 073763-5G
102531-870EA 153.85 USD
102531-870 102531-872
7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥95%
7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MW: 214.65 g/mol
MDL Number: MFCD07371494
CAS Number: 99365-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR