3,4-Dihydropyrrolo[1,2-a]pyrazin-1(2H)-one ≥95%

Supplier: Matrix Scientific

073762-250MG 073762-1G 073762-5G
102531-864EA 1044.62 USD
102531-864 102531-866 102531-868
3,4-Dihydropyrrolo[1,2-a]pyrazin-1(2H)-one ≥95%
3,4-Dihydropyrrolo[1,2-a]pyrazin-1(2H)-one

Hazard: Irritant

Formula: C₇H₈N₂O
MDL Number: MFCD09832035
CAS Number: 54906-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR