Methyl-3-methyl-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-methyl-1H-pyrazole-4-carboxylate

073761-250MG 073761-1G 073761-5G
102531-858EA 283.08 USD
102531-858 102531-860 102531-862
Methyl-3-methyl-1H-pyrazole-4-carboxylate ≥95%
Methyl-3-methyl-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₆H₈N₂O₂
MW: 140.1400 g/mol
MDL Number: MFCD07357354
CAS Number: 23170-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR