2,7-Dichlorothiazolo[5,4-c]pyridine ≥95%

Supplier: Matrix Scientific

073758-250MG 073758-1G
102531-842EA 1223.08 USD
102531-842 102531-844
2,7-Dichlorothiazolo[5,4-c]pyridine ≥95%
2,7-Dichlorothiazolo[5,4-c]pyridine

Hazard: Irritant

Formula: C₆H₂Cl₂N₂S
MW: 205.0600 g/mol
MDL Number: MFCD13193435
CAS Number: 884860-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR